missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-4-nitropyridine N-oxide, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC308990050

 View more versions of this product

Catalog No. AC308990050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Crystalline Needles or Powder
154.13
Yellow
100.0
98%
MFCD00014626
Glass bottle
3-Methyl-4-nitropyridine N-oxide
≥97.5%
5g
135° to 139°C
1074-98-2
C6H6N2O3
97.5
Authentic

Chemical Identifiers

1074-98-2
100.0
C6H6N2O3
MFCD00014626
≥97.5%
97.5
98%
154.13
3-Methyl-4-nitropyridine N-oxide

Safety and Handling

SDS & Safety Documents

GHS P Statement
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning