missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-4-nitropyrazole 98.0+%, TCI America™

$123.88 - $413.42

Chemical Identifiers

CAS 5334-39-4
Molecular Formula C4H5N3O2
Molecular Weight (g/mol) 127.103
MDL Number MFCD00037864
InChI Key WTZYTQJELOHMMJ-UHFFFAOYSA-N
Synonym 1h-pyrazole, 3-methyl-4-nitro, 3-me-4-no2 pyrazole, 3-methyl-4-nitro pyrazole, 3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole #, 3-methyl-4-nitropyrazole, 4-nitro-5-methyl-1h-pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j
PubChem CID 79255
IUPAC Name 5-methyl-4-nitro-1H-pyrazole
SMILES CC1=C(C=NN1)[N+](=O)[O-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M24695G
View Documents
Tci America
M24695G
5g Each for $123.88
 
M246925G
View Documents
Tci America
M246925G
25g Each for $413.42
 
Specifications

Chemical Identifiers

5334-39-4
127.103
WTZYTQJELOHMMJ-UHFFFAOYSA-N
79255
CC1=C(C=NN1)[N+](=O)[O-]
C4H5N3O2
MFCD00037864
1h-pyrazole, 3-methyl-4-nitro, 3-me-4-no2 pyrazole, 3-methyl-4-nitro pyrazole, 3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole #, 3-methyl-4-nitropyrazole, 4-nitro-5-methyl-1h-pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j
5-methyl-4-nitro-1H-pyrazole

Specifications

325°C
135°C
5334-39-4
MFCD00037864
WTZYTQJELOHMMJ-UHFFFAOYSA-N
5-methyl-4-nitro-1H-pyrazole
79255
Crystal-Powder at 20°C
White-Yellow
5g
C4H5N3O2
1h-pyrazole, 3-methyl-4-nitro, 3-me-4-no2 pyrazole, 3-methyl-4-nitro pyrazole, 3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole #, 3-methyl-4-nitropyrazole, 4-nitro-5-methyl-1h-pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j
CC1=C(C=NN1)[N+](=O)[O-]
127.103
127.10
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

RTECSNumber : UQ7435000

TSCA : No