missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-4-nitropyrazole 98.0+%, TCI America™

Manufacturer:  Tci America M246925G

 View more versions of this product

Catalog No. M246925G



Specifications

Specifications

3-Methyl-4-nitropyrazole
White-Yellow
25g
C4H5N3O2
1h-pyrazole, 3-methyl-4-nitro, 3-me-4-no2 pyrazole, 3-methyl-4-nitro pyrazole, 3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole #, 3-methyl-4-nitropyrazole, 4-nitro-5-methyl-1h-pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j
CC1=C(C=NN1)[N+](=O)[O-]
127.103
127.10
≥98.0% (GC,T)
325°C
135°C
5334-39-4
MFCD00037864
WTZYTQJELOHMMJ-UHFFFAOYSA-N
5-methyl-4-nitro-1H-pyrazole
79255
Crystal-Powder at 20°C

Chemical Identifiers

5334-39-4
127.103
WTZYTQJELOHMMJ-UHFFFAOYSA-N
79255
CC1=C(C=NN1)[N+](=O)[O-]
C4H5N3O2
MFCD00037864
1h-pyrazole, 3-methyl-4-nitro, 3-me-4-no2 pyrazole, 3-methyl-4-nitro pyrazole, 3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole #, 3-methyl-4-nitropyrazole, 4-nitro-5-methyl-1h-pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j
5-methyl-4-nitro-1H-pyrazole
Safety and Handling

Safety and Handling

RTECSNumber : UQ7435000

TSCA : No

Documents
Promotions

Promotions