Please login to your online account to display your discounted pricing

3-Methyl-4-nitrophenol 98%, ACROS Organics™


Quantity 500g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  127305000

 View more versions of this product

Catalog No. AC127305000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 153.14
Color Yellow to beige
Quantity 500g
RTECS Number GP2625000
EINECS Number 219-952-5
TSCA TSCA
Melting Point 126° to 130°C
Beilstein 06, 386
CAS Max % 100.0
Synonym 4-Nitro-m-cresol
Infrared Spectrum Authentic
Health Hazard 1 GHS Signal Word: Warning
CAS Min % 97.5
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
CAS 2581-34-2
Molecular Formula C7H7NO3
Chemical Name or Material 3-Methyl-4-nitrophenol
Name Note 98%
Linear Formula CH3C6H3(NO2)OH
MDL Number MFCD00007333
Assay Percent Range 98%

Specifications

Chemical Name or Material 3-Methyl-4-nitrophenol
Name Note 98%
CAS 2581-34-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7NO3
Linear Formula CH3C6H3(NO2)OH
Formula Weight 153.14
MDL Number MFCD00007333
Synonym 4-Nitro-m-cresol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents