Please login to your online account to display your discounted pricing

3-Methyl-4-nitrophenol 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methyl-4-nitrophenol
Name Note 98%
CAS 2581-34-2
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C7H7NO3
Linear Formula CH3C6H3(NO2)OH
Formula Weight 153.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC127305000

  SDS
thermo fisher scientific
127305000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Crystalline Powder
Formula Weight 153.14
Color Yellow to beige
Quantity 500g
RTECS Number GP2625000
EINECS Number 219-952-5
TSCA TSCA
Melting Point 126° to 130°C
Beilstein 06, 386
CAS Max % 100.0
Infrared Spectrum Authentic
Synonym 4-Nitro-m-cresol
Chemical Name or Material 3-Methyl-4-nitrophenol
Name Note 98%
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C7H7NO3
Linear Formula CH3C6H3(NO2)OH
MDL Number MFCD00007333
CAS 2581-34-2
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.