Please login to your online account to display your discounted pricing

3-Methyl-4-nitrobenzyl alcohol, 98%, ACROS Organics™

$32.47 - $32.47

Chemical Identifiers

Chemical Name or Material 3-Methyl-4-nitrobenzyl alcohol
CAS 80866-75-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9NO3
Linear Formula CH3C6H3(NO2)CH2OH
Formula Weight 167.16
MDL Number MFCD00007171
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC182100010

  SDS
thermo fisher scientific
182100010
Glass bottle 1g Each for $32.47
Description & Specifications

Specifications

Assay ≥97.5%
Formula Weight 167.16
Physical Form Powder
Quantity 1g
Color Yellow to ochre
Melting Point 56° to 60°C
CAS Max % 100.0
CAS 80866-75-7
Chemical Name or Material 3-Methyl-4-nitrobenzyl alcohol
Molecular Formula C8H9NO3
MDL Number MFCD00007171
CAS Min % 97.5
Infrared Spectrum Authentic
Packaging Glass bottle
Linear Formula CH3C6H3(NO2)CH2OH
Assay Percent Range 98%

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.