Please login to your online account to display your discounted pricing

3-Methyl-4-nitrobenzyl alcohol, 98%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  182100010

 View more versions of this product

Catalog No. AC182100010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 167.16
Quantity 1g
Color Yellow to ochre
Melting Point 56° to 60°C
CAS Max % 100.0
Chemical Name or Material 3-Methyl-4-nitrobenzyl alcohol
Infrared Spectrum Authentic
Packaging Glass bottle
Molecular Formula C8H9NO3
Linear Formula CH3C6H3(NO2)CH2OH
MDL Number MFCD00007171
CAS 80866-75-7
CAS Min % 97.5
Assay Percent Range 98%

Specifications

Chemical Name or Material 3-Methyl-4-nitrobenzyl alcohol
CAS 80866-75-7
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9NO3
Linear Formula CH3C6H3(NO2)CH2OH
Formula Weight 167.16
MDL Number MFCD00007171