missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-4-nitrobenzoic acid, 99%

Manufacturer:  Alfa Aesar™ A1552322

 View more versions of this product

Catalog No. AAA1552322



Specifications

Specifications

3-Methyl-4-nitrobenzoic acid
100g
C8H7NO4
1959154
XDTTUTIFWDAMIX-UHFFFAOYSA-N
181.147
181.15
215°C to 220°C
3113-71-1
MFCD00007168
3-methyl-4-nitrobenzoic acid, 4-nitro-m-toluic acid, benzoic acid, 3-methyl-4-nitro, m-toluic acid, 4-nitro, unii-elp258jxh0, 3-methyl-4-nitrobenzoicacid, 3-methyl-4-nitro-benzoic acid, elp258jxh0, benzoic acid,3-methyl-4-nitro, 3-methyl-4-nitro benzoic acid
CC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]
18370
99%

Chemical Identifiers

3113-71-1
181.147
XDTTUTIFWDAMIX-UHFFFAOYSA-N
18370
C8H7NO4
MFCD00007168
3-methyl-4-nitrobenzoic acid, 4-nitro-m-toluic acid, benzoic acid, 3-methyl-4-nitro, m-toluic acid, 4-nitro, unii-elp258jxh0, 3-methyl-4-nitrobenzoicacid, 3-methyl-4-nitro-benzoic acid, elp258jxh0, benzoic acid,3-methyl-4-nitro, 3-methyl-4-nitro benzoic acid
CC1=C(C=CC(=C1)C(=O)O)[N+](=O)[O-]
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents