Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-4-nitrobenzoic acid, 99%

$61.26 - $261.68

Chemical Identifiers

Chemical Name or Material 3-Methyl-4-nitrobenzoic acid
CAS 3113-71-1
Molecular Formula C8H7NO4
Formula Weight 181.15
MDL Number MFCD00007168
Beilstein 1959154
Synonym 4-Nitro-m-toluic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1552322

 
alfa aesar™
A1552322
100g Each for $61.26

AAA1552336

 
alfa aesar™
A1552336
500g Each for $261.68
Description & Specifications

Specifications

Formula Weight 181.15
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 3-Methyl-4-nitrobenzoic acid
TSCA TSCA
Assay 99%
Molecular Formula C8H7NO4
MDL Number MFCD00007168
CAS 3113-71-1
Beilstein 1959154
Synonym 4-Nitro-m-toluic acid
Melting Point 215° to 220°C
Quantity 100g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.