Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-4-nitrobenzamide, 97%

$78.04 - $826.59

Chemical Identifiers

Chemical Name or Material 3-Methyl-4-nitrobenzamide
CAS 99584-85-7
Molecular Formula C8H8N2O3
Formula Weight 180.16
MDL Number MFCD00031468
Beilstein 2264048
Synonym 4-Nitro-m-toluamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2469103

 
alfa aesar™
B2469103
1g Each for $78.04

AAB2469106

 
alfa aesar™
B2469106
5g Each for $204.73

AAB2469114

 
alfa aesar™
B2469114
25g Each for $826.59
Description & Specifications

Specifications

Formula Weight 180.16
Chemical Name or Material 3-Methyl-4-nitrobenzamide
Assay 97%
Molecular Formula C8H8N2O3
MDL Number MFCD00031468
CAS 99584-85-7
Beilstein 2264048
Synonym 4-Nitro-m-toluamide
Health Hazard 1 Danger
Melting Point 158° to 160°C
Quantity 1g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Applications
3-Methyl-4-nitrobenzamide is used as synthetic organic intermediates.

Solubility
Soluble in alcohol, slightly soluble in hot water.

Notes
Store in a cool, dry place in tightly closed container. Incompatible with oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.