Please login to your online account to display your discounted pricing

3-Methyl-4-nitroanisole, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: fisher scientific acros  166530050

 View more versions of this product

Catalog No. AC166530050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 167.16
Quantity 5g
Color Yellow to brown
Beilstein 06, 386
EINECS Number 226-356-9
TSCA TSCA
Melting Point 48° to 52°C
CAS Max % 100.0
Chemical Name or Material 3-Methyl-4-nitroanisole
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H9NO3
Linear Formula CH3C6H3(NO2)OCH3
MDL Number MFCD00007167
CAS 5367-32-8
CAS Min % 97.5

Specifications

Chemical Name or Material 3-Methyl-4-nitroanisole
Name Note 98%
CAS 5367-32-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9NO3
Linear Formula CH3C6H3(NO2)OCH3
Formula Weight 167.16
MDL Number MFCD00007167