Please login to your online account to display your discounted pricing

3-Methyl-4-nitroanisole, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methyl-4-nitroanisole
Name Note 98%
CAS 5367-32-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H9NO3
Linear Formula CH3C6H3(NO2)OCH3
Formula Weight 167.16
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC166531000

  SDS
fisher scientific acros
166531000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC166530250

  SDS
fisher scientific acros
166530250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 167.16
Quantity 100g
Color Yellow to brown
Beilstein 06, 386
EINECS Number 226-356-9
TSCA TSCA
Melting Point 48° to 52°C
CAS Max % 100.0
CAS Min % 97.5
Chemical Name or Material 3-Methyl-4-nitroanisole
Infrared Spectrum Authentic
Packaging Glass bottle
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H9NO3
Linear Formula CH3C6H3(NO2)OCH3
MDL Number MFCD00007167
CAS 5367-32-8