Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-4-nitroanisole, 97%

$46.41 - $302.86

Chemical Identifiers

Chemical Name or Material 3-Methyl-4-nitroanisole
CAS 5367-32-8
Molecular Formula C8H9NO3
Formula Weight 167.16
MDL Number MFCD00007167
Beilstein 2328600
Synonym 5-Methoxy-2-nitrotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2234814

  SDS
alfa aesar™
B2234814
25g Each for $118.28

AAB2234806

  SDS
alfa aesar™
B2234806
5g Each for $46.41

AAB2234822

  SDS
alfa aesar™
B2234822
100g Each for $302.86
Description & Specifications

Specifications

Formula Weight 167.16
Molecular Formula C8H9NO3
CAS 5367-32-8
Synonym 5-Methoxy-2-nitrotoluene
Quantity 25g
Melting Point 48° to 50°C
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Beilstein 2328600
MDL Number MFCD00007167
Assay 97%
Chemical Name or Material 3-Methyl-4-nitroanisole
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Danger
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.