Learn More
3-Methyl-4-nitro-1H-pyrazole, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals H3086014
Description
3-Methyl-4-nitropyrazole is a reactant used in the preparation of aminopyrazole derivatives as potent, selective and brain penetrant Leucine-rich repeat kinase 2(LRRK2) small molecule inhibitors.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methyl-4-nitropyrazole is a reactant used in the preparation of aminopyrazole derivatives as potent, selective and brain penetrant Leucine-rich repeat kinase 2(LRRK2) small molecule inhibitors.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
3-Methyl-4-nitro-1H-pyrazole | |
131°C to 134°C | |
C4H5N3O2 | |
25 g | |
Slightly soluble in water. | |
CC1=C(C=NN1)[N+]([O-])=O | |
127.10 | |
127.1 |
5334-39-4 | |
325°C | |
MFCD00037864 | |
3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitropyrazole, 1h-pyrazole, 3-methyl-4-nitro, 3-methyl-4-nitro pyrazole, 3-me-4-no2 pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j, 4-nitro-5-methyl-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole # | |
WTZYTQJELOHMMJ-UHFFFAOYSA-N | |
5-methyl-4-nitro-1H-pyrazole | |
79255 | |
97% |
Chemical Identifiers
5334-39-4 | |
127.10 | |
WTZYTQJELOHMMJ-UHFFFAOYSA-N | |
79255 | |
CC1=C(C=NN1)[N+]([O-])=O |
C4H5N3O2 | |
MFCD00037864 | |
3-methyl-4-nitro-1h-pyrazole, 3-methyl-4-nitropyrazole, 1h-pyrazole, 3-methyl-4-nitro, 3-methyl-4-nitro pyrazole, 3-me-4-no2 pyrazole, 5-methyl-4-nitropyrazole, acmc-209l6a, ksc497o4j, 4-nitro-5-methyl-1h-pyrazole, 3-methyl-4-nitro-1h-pyrazole # | |
5-methyl-4-nitro-1H-pyrazole |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
RTECSNumber : UQ7435000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only