Please login to your online account to display your discounted pricing

3-methyl-4-isoxazolecarboxylic acid, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 3-Methyl-4-isoxazolecarboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5NO3
Formula Weight 127.1
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

MO01252CB

  SDS
fisher scientific company
MO01252CB
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 127.1
Quantity 250mg
Assay 97%
Chemical Name or Material 3-Methyl-4-isoxazolecarboxylic acid
Assay Percent Range 97%
Molecular Formula C5H5NO3
CAS 17153-20-7