Please login to your online account to display your discounted pricing

3-methyl-4-isoxazolecarboxylic acid, 97%, Maybridge


Quantity 250mg

Manufacturer: fisher scientific company  MO01252CB

 View more versions of this product

Catalog No. MO01252CB

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 127.1
Quantity 250mg
Assay 97%
Chemical Name or Material 3-Methyl-4-isoxazolecarboxylic acid
CAS 17153-20-7
Molecular Formula C5H5NO3
Assay Percent Range 97%

Specifications

Chemical Name or Material 3-Methyl-4-isoxazolecarboxylic acid
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H5NO3
Formula Weight 127.1