Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-4-decanol, erythro + threo, 97%

$38.95 - $606.47

Chemical Identifiers

Chemical Name or Material 3-Methyl-4-decanol
CAS 55816-17-6
Name Note erythro + threo
Molecular Formula C11H24O
Formula Weight 172.31
MDL Number MFCD00046702
Synonym 2-Butyl n-hexyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2005003

 
alfa aesar™
B2005003
1g Each for $38.95

AAB2005014

 
alfa aesar™
B2005014
25g Each for $606.47
Description & Specifications

Specifications

Formula Weight 172.31
Chemical Name or Material 3-Methyl-4-decanol
Name Note erythro + threo
Assay 97%
Molecular Formula C11H24O
MDL Number MFCD00046702
CAS 55816-17-6
Synonym 2-Butyl n-hexyl carbinol
Health Hazard 1 Warning
Quantity 1g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.