missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-3-trimethylsiloxy-1-butyne, 97%

Manufacturer:  Alfa Aesar™ 4392209

 View more versions of this product

Catalog No. AA4392209



Specifications

Specifications

3-Methyl-3-trimethylsiloxy-1-butyne
1.4052
10g
C8H16OSi
UN1993
JNRUXZIXAXHXTN-UHFFFAOYSA-N
trimethyl(2-methylbut-3-yn-2-yloxy)silane
87344
Liquid
Characteristic
52°C (75mmHg)
17869-77-1
MFCD00053867
3-methyl-3-trimethylsilyloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yloxy silane, trimethyl 2-methylbut-3-yn-2-yl oxy silane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 3-methyl-3-trimethylsiloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yl oxy silane, acmc-1brai, silane, 1,1-dimethyl-2-propynyl oxy trimethyl, 3-trimethylsiloxy-3-methyl-1-butyne
CC(C)(C#C)O[Si](C)(C)C
156.3
156.3
97%

Chemical Identifiers

17869-77-1
156.3
JNRUXZIXAXHXTN-UHFFFAOYSA-N
87344
CC(C)(C#C)O[Si](C)(C)C
C8H16OSi
MFCD00053867
3-methyl-3-trimethylsilyloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yloxy silane, trimethyl 2-methylbut-3-yn-2-yl oxy silane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 1,1-dimethyl-2-propynyl oxy trimethylsilane, 3-methyl-3-trimethylsiloxy-1-butyne, trimethyl 2-methylbut-3-yn-2-yl oxy silane, acmc-1brai, silane, 1,1-dimethyl-2-propynyl oxy trimethyl, 3-trimethylsiloxy-3-methyl-1-butyne
trimethyl(2-methylbut-3-yn-2-yloxy)silane
Safety and Handling

Safety and Handling

GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents