Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-3-penten-2-one, (E)+(Z), 95%

$22.88 - $22.88

Chemical Identifiers

Chemical Name or Material 3-Methyl-3-penten-2-one
CAS 565-62-8
Name Note (E)+(Z)
Molecular Formula C6H10O
Formula Weight 98.15
MDL Number MFCD00151839
UN Number UN1224
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1354809

 
alfa aesar™
L1354809
10g Each for $22.88

AAL1354818

 
alfa aesar™
L1354818
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 98.15
Chemical Name or Material 3-Methyl-3-penten-2-one
TSCA TSCA
Assay 95%
Molecular Formula C6H10O
CAS 565-62-8
Health Hazard 1 Warning
Melting Point −70°C
Boiling Point 138°C
Quantity 10g
Name Note (E)+(Z)
MDL Number MFCD00151839
UN Number UN1224
Flash Point 34°C (93°F)
Health Hazard 2 GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.