missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-3-penten-2-one, (E)+(Z), 95%

Manufacturer:  Alfa Aesar™ L1354809

 View more versions of this product

Catalog No. AAL1354809



Specifications

Specifications

3-Methyl-3-penten-2-one
138°C
10g
565-62-8
MFCD00151839
3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene
CC=C(C)C(=O)C
98.145
98.15
34°C (93°F)
-70°C
(E)+(Z)
C6H10O
UN1224
ZAMCMCQRTZKGDX-SNAWJCMRSA-N
(E)-3-methylpent-3-en-2-one
5364579
95%

Chemical Identifiers

565-62-8
98.145
ZAMCMCQRTZKGDX-SNAWJCMRSA-N
5364579
CC=C(C)C(=O)C
C6H10O
MFCD00151839
3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene
(E)-3-methylpent-3-en-2-one
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

TSCA : TSCA

Documents