missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-3-penten-2-one, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 438440500

 View more versions of this product

Catalog No. AC438440500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

3-Methyl-3-penten-2-one
97.5
0.8750g/mL
34°C
97.5% min. (GC)
C6H10O
50g
3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene
ZAMCMCQRTZKGDX-SNAWJCMRSA-N
(E)-3-methylpent-3-en-2-one
5364579
98%
565-62-8
100.0
136°C to 139°C
Authentic
Glass bottle
CH3CH=C(CH3)COCH3
0.875
Solubility in water: 29.1g/L (20°C)
CC=C(C)C(=O)C
98.14
98.14

Chemical Identifiers

565-62-8
98.14
3-methyl-3-penten-2-one, e-3-methylpent-3-en-2-one, e-3-methyl-3-penten-2-one, unii-jbr4o01db2, jbr4o01db2, 3-methyl-2-penten-4-one, 3-penten-2-one, 3-methyl, 3-methyl-3-pentene-2-one, 2-acetyl-2-butene, 3-methyl-4-oxopent-2-ene
(E)-3-methylpent-3-en-2-one
C6H10O
ZAMCMCQRTZKGDX-SNAWJCMRSA-N
5364579
CC=C(C)C(=O)C
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a P

GHS Signal Word: Warning

EINECSNumber : 209-283-7

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only