missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-3-oxetanemethanol, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC240840500

 View more versions of this product

Catalog No. AC240840500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

3-Methyl-3-oxetanemethanol
97°C
Glass bottle
122.0°C (70.0 mmHg)
50g
97.5
97.5% min. (GC)
MFCD00010273
NLQMSBJFLQPLIJ-UHFFFAOYSA-N
(3-methyloxetan-3-yl)methanol
137837
Liquid
1.0340g/mL
Authentic
1.034
Colorless to Yellow
3143-02-0
100.0
C5H10O2
3-hydroxymethyl-3-methyl-oxetane, 3-hydroxymethyl-3-methyloxetane, 3-methyl-3-hydroxymethyloxetane, 3-methyl-3-oxetanemethanol, 3-methyl-3-oxetanyl methanol, 3-methyl-3-oxethanemethanol, 3-methyl-oxetan-3-yl-methanol, 3-methyloxetan-3-yl methanol, 3-methyloxetane-3-methanol, 3-oxetanemethanol, 3-methyl
CC1(COC1)CO
102.13
102.13
98%

Chemical Identifiers

3143-02-0
102.13
NLQMSBJFLQPLIJ-UHFFFAOYSA-N
137837
CC1(COC1)CO
C5H10O2
MFCD00010273
3-hydroxymethyl-3-methyl-oxetane, 3-hydroxymethyl-3-methyloxetane, 3-methyl-3-hydroxymethyloxetane, 3-methyl-3-oxetanemethanol, 3-methyl-3-oxetanyl methanol, 3-methyl-3-oxethanemethanol, 3-methyl-oxetan-3-yl-methanol, 3-methyloxetan-3-yl methanol, 3-methyloxetane-3-methanol, 3-oxetanemethanol, 3-methyl
(3-methyloxetan-3-yl)methanol
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If s

GHS Signal Word: Warning

SDS Documents