missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-2-thiophenecarboxaldehyde, 90%, Tech., ACROS Organics™

Chemical Identifiers

CAS 5834-16-2
CAS Min % 88.0
CAS Max % 100.0
Assay Percent Range 90%
Molecular Formula C6H6OS
Formula Weight 126.17
MDL Number MFCD00005430
Chemical Name or Material 3-Methyl-2-thiophenecarboxaldehyde, tech. 90%
Assay ≥88%
Grade Technical Grade
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC172781000 SDS Thermo Fisher Scientific
172781000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

5834-16-2
100.0
C6H6OS
MFCD00005430
≥88%
88.0
90%
126.17
3-Methyl-2-thiophenecarboxaldehyde, tech. 90%
Technical Grade

Specifications

Clear yellow to brown
5834-16-2
100.0
Authentic
Liquid
C6H6OS
MFCD00005430
3-Methyl-2-thiophenecarboxaldehyde, tech. 90%
Technical Grade
100g
88.0
1.1700g/mL
Glass bottle
90%
126.17
17, III, 4521
≥88%

Safety and Handling

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 227-418-8

TSCA : TSCA