Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-2-thiohydantoin, 95%

Chemical Identifiers

Chemical Name or Material 3-Methyl-2-thiohydantoin
CAS 694-68-8
Formula Weight 130.16
MDL Number MFCD00038425
Beilstein 2840
Synonym 3-Methyl-2-thioxo-4-imidazolidinone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6315703

 
alfa aesar™
H6315703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6315706

 
alfa aesar™
H6315706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63157MD

 
alfa aesar™
H63157MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 130.16
Chemical Name or Material 3-Methyl-2-thiohydantoin
Physical Form Solid
Assay 95%
MDL Number MFCD00038425
CAS 694-68-8
Beilstein 2840
Synonym 3-Methyl-2-thioxo-4-imidazolidinone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Quantity 1g
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.