Please login to your online account to display your discounted pricing

(3-Methyl-2-thienyl)methylamine, 97%, Maybridge

$283.95 - $283.95

Chemical Identifiers

Chemical Name or Material (3-Methyl-2-thienyl)methylamine
CAS 104163-35-1
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C6H9NS
Formula Weight 127.21
MDL Number MFCD02093988
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SP01443DA

 
fisher scientific company
SP01443DA
Amber Glass Bottle 1g Each for $283.95
Description & Specifications

Specifications

Formula Weight 127.21
Quantity 1g
CAS Min % 97.0
Packaging Amber Glass Bottle
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Chemical Name or Material (3-Methyl-2-thienyl)methylamine
Assay Percent Range 97%
MDL Number MFCD02093988
CAS 104163-35-1
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Molecular Formula C6H9NS
Safety and Handling

Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.