missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-2-pyrazolin-5-one, 98+%

$52.12 - $658.49

Chemical Identifiers

CAS 108-26-9
Molecular Formula C4H6N2O
Molecular Weight (g/mol) 98.105
MDL Number MFCD00020699
InChI Key NHLAPJMCARJFOG-UHFFFAOYSA-N
Synonym 3-methyl-2-pyrazolin-5-one, 3-methyl-5-pyrazolone, 3-methyl-1h-pyrazol-5 4h-one, 3h-pyrazol-3-one, 2,4-dihydro-5-methyl, 3-methyl-pyrazolon-5, 2-pyrazolin-5-one, 3-methyl, 3-methyl-4,5-dihydro-1h-pyrazol-5-one, 5-methyl-2,4-dihydro-3h-pyrazol-3-one, 2,4-dihydro-5-methyl-3h-pyrazol-3-one, 3-methyl-pyrazolon-5 german
PubChem CID 7920
IUPAC Name 3-methyl-1,4-dihydropyrazol-5-one
SMILES CC1=NNC(=O)C1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1025318 View Documents Alfa Aesar™
A1025318
50g Each for $189.68
 
AAA1025309 View Documents Alfa Aesar™
A1025309
10g Each for $52.12
 
AAA1025330 View Documents Alfa Aesar™
A1025330
250g Each for $658.49
 
Description

Description

Applications
3-Methyl-2-pyrazolin-5-one is used in the preparation of spirocyclic [indole-3,4′-pyrano[2,3-c]pyrazole] by reacting with isatins and malonitrile in the presence of sodium bromide as electrolyte.

Solubility
Slightly soluble in ethanol.

Notes
Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

108-26-9
98.105
NHLAPJMCARJFOG-UHFFFAOYSA-N
7920
CC1=NNC(=O)C1
C4H6N2O
MFCD00020699
3-methyl-2-pyrazolin-5-one, 3-methyl-5-pyrazolone, 3-methyl-1h-pyrazol-5 4h-one, 3h-pyrazol-3-one, 2,4-dihydro-5-methyl, 3-methyl-pyrazolon-5, 2-pyrazolin-5-one, 3-methyl, 3-methyl-4,5-dihydro-1h-pyrazol-5-one, 5-methyl-2,4-dihydro-3h-pyrazol-3-one, 2,4-dihydro-5-methyl-3h-pyrazol-3-one, 3-methyl-pyrazolon-5 german
3-methyl-1,4-dihydropyrazol-5-one

Specifications

220°C to 224°C
108-26-9
MFCD00020699
NHLAPJMCARJFOG-UHFFFAOYSA-N
3-methyl-1,4-dihydropyrazol-5-one
7920
≥98%
50g
C4H6N2O
3-methyl-2-pyrazolin-5-one, 3-methyl-5-pyrazolone, 3-methyl-1h-pyrazol-5 4h-one, 3h-pyrazol-3-one, 2,4-dihydro-5-methyl, 3-methyl-pyrazolon-5, 2-pyrazolin-5-one, 3-methyl, 3-methyl-4,5-dihydro-1h-pyrazol-5-one, 5-methyl-2,4-dihydro-3h-pyrazol-3-one, 2,4-dihydro-5-methyl-3h-pyrazol-3-one, 3-methyl-pyrazolon-5 german
CC1=NNC(=O)C1
98.105
98.11
3-Methyl-2-pyrazolin-5-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : UQ9451500

TSCA : TSCA