missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
3-Methyl-2-pentene (cis- and trans- mixture) 99.0+%, TCI America™
$114.48 - $114.48
Chemical Identifiers
CAS | 922-61-2 |
---|---|
Molecular Formula | C6H12 |
Molecular Weight (g/mol) | 84.162 |
MDL Number | MFCD00009334 |
InChI Key | BEQGRRJLJLVQAQ-XQRVVYSFSA-N |
PubChem CID | 643935 |
IUPAC Name | (Z)-3-methylpent-2-ene |
SMILES | CCC(=CC)C |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
M03151ML
|
TCI America
M03151ML |
1 mL |
Each for $114.48
|
|
|||||
Chemical Identifiers
922-61-2 | |
84.162 | |
BEQGRRJLJLVQAQ-XQRVVYSFSA-N | |
(Z)-3-methylpent-2-ene |
C6H12 | |
MFCD00009334 | |
643935 | |
CCC(=CC)C |
Specifications
922-61-2 | |
C6H12 | |
1 mL | |
BEQGRRJLJLVQAQ-XQRVVYSFSA-N | |
(Z)-3-methylpent-2-ene | |
643935 | |
≥99.0% (GC) | |
3-Methyl-2-pentene (cis- and trans- mixture) |
Colorless | |
MFCD00009334 | |
2288 | |
CCC(=CC)C | |
84.162 | |
84.16 | |
Liquid |
Safety and Handling
EINECSNumber : (2)-0031
TSCA : No