Please login to your online account to display your discounted pricing

3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-carboxaldehyde, 97%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  432690010

 View more versions of this product

Catalog No. AC432690010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Melting Point 146° to 150°C
Color White to off-white
Quantity 1g
Chemical Name or Material 3-Methyl-2-oxo-2, 3-dihydro-1, 3-benzoxazole-6-carboxaldehyde
CAS 54903-66-1
CAS Min % 96.0
CAS Max % 100.0
Physical Form White to off-white powder
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C9H7NO3
Formula Weight 177.16g/mol
MDL Number MFCD17171428
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 3-Methyl-2-oxo-2, 3-dihydro-1, 3-benzoxazole-6-carboxaldehyde
CAS 54903-66-1
CAS Min % 96.0
CAS Max % 100.0
Assay 96.0% min.
Assay Percent Range 97%
Molecular Formula C9H7NO3
Formula Weight 177.16g/mol
MDL Number MFCD17171428
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents