Please login to your online account to display your discounted pricing

3-Methyl-2-nitrophenol, 97%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  171090050

 View more versions of this product

Catalog No. AC171090050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Formula Weight 153.14
Physical Form Crystalline Powder or Low Melting Mass
Quantity 5g
Color Yellow to orange-brown
TSCA TSCA
Melting Point 36° to 41°C
EINECS Number 225-546-9
Beilstein 06, 385
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Synonym 2-Nitro-m-cresol
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Chemical Name or Material 3-Methyl-2-nitrophenol
Molecular Formula C7H7NO3
Linear Formula CH3C6H3(NO2)OH
CAS 4920-77-8
MDL Number MFCD00007125
Assay Percent Range 97%

Specifications

Chemical Name or Material 3-Methyl-2-nitrophenol
Name Note 97%
CAS 4920-77-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H7NO3
Linear Formula CH3C6H3(NO2)OH
Formula Weight 153.14
MDL Number MFCD00007125
Synonym 2-Nitro-m-cresol
Accessories

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

SDS & Safety Documents