Please login to your online account to display your discounted pricing

3-Methyl-2-nitrophenol, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methyl-2-nitrophenol
Name Note 97%
CAS 4920-77-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H7NO3
Linear Formula CH3C6H3(NO2)OH
Formula Weight 153.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC171090050

  SDS
thermo fisher scientific
171090050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 153.14
Physical Form Crystalline Powder or Low Melting Mass
Quantity 5g
Color Yellow to orange-brown
TSCA TSCA
Melting Point 36° to 41°C
EINECS Number 225-546-9
Beilstein 06, 385
CAS Min % 96.0
CAS Max % 100.0
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Warning
Infrared Spectrum Authentic
Synonym 2-Nitro-m-cresol
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 97%
Chemical Name or Material 3-Methyl-2-nitrophenol
Molecular Formula C7H7NO3
Linear Formula CH3C6H3(NO2)OH
CAS 4920-77-8
MDL Number MFCD00007125
Assay Percent Range 97%
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.