Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-2-nitroanisole, 98%

$25.54 - $355.57

Chemical Identifiers

Chemical Name or Material 3-Methyl-2-nitroanisole
CAS 5345-42-6
Molecular Formula C8H9NO3
Formula Weight 167.16
MDL Number MFCD00007179
Beilstein 2329690
Synonym 2-Methoxy-6-methylnitrobenzene; 3-Methoxy-2-nitrotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1889014

 
alfa aesar™
A1889014
25g Each for $355.57

AAA1889003

 
alfa aesar™
A1889003
1g Each for $25.54

AAA1889006

 
alfa aesar™
A1889006
5g Each for $84.20
Description & Specifications

Specifications

Formula Weight 167.16
Chemical Name or Material 3-Methyl-2-nitroanisole
Assay 98%
Molecular Formula C8H9NO3
MDL Number MFCD00007179
CAS 5345-42-6
Beilstein 2329690
Synonym 2-Methoxy-6-methylnitrobenzene; 3-Methoxy-2-nitrotoluene
Health Hazard 1 Danger
Melting Point 48° to 52°C
Quantity 25g
Flash Point >110°C (230°F)
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.