Please login to your online account to display your discounted pricing

3-Methyl-2-cyclopenten-1-one, 98%, stabilized , ACROS Organics™

$133.61 - $133.61

Chemical Identifiers

Chemical Name or Material 3-Methyl-2-cyclopenten-1-one
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C6H8O
Linear Formula CH3C5H5(=O)
Formula Weight 96.13
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC163590100

  SDS
fisher scientific acros
163590100
Glass bottle 10g Each for $133.61

AC163590500

  SDS
fisher scientific acros
163590500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC163592500

  SDS
thermo fisher scientific
AC163592500
Glass bottle 250g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (GC)
Quantity 10g
Physical Form Liquid
Formula Weight 96.13
Color Clear light yellow to yellow-brown
Density 0.9800g/mL
Packaging Glass bottle
Assay Percent Range 98%
Molecular Formula C6H8O
Infrared Spectrum Authentic
Stabilizer 0.1% hydroquinone
CAS 2758-18-1
Refractive Index 1.4870 to 1.4900 (20°C, 589nm)
Chemical Name or Material 3-Methyl-2-cyclopenten-1-one
Linear Formula CH3C5H5(=O)
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes serious eye irritation.