missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-2-cyclohexen-1-one, TCI America™

Manufacturer:  TCI America M104325G

 View more versions of this product

Catalog No. M104325G



Specifications

Specifications

3-Methyl-2-cyclohexen-1-one
Yellow
1193-18-6
MFCD00001581
IITQJMYAYSNIMI-UHFFFAOYSA-N
3-methylcyclohex-2-en-1-one
14511
Clear Liquid at 20°C
197°C
25g
C7H10O
3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
CC1=CC(=O)CCC1
110.156
110.16
≥98.0% (GC)

Chemical Identifiers

1193-18-6
110.156
IITQJMYAYSNIMI-UHFFFAOYSA-N
14511
CC1=CC(=O)CCC1
C7H10O
MFCD00001581
3-methyl-2-cyclohexen-1-one, seudenone, 3-methyl-2-cyclohexenone, 2-cyclohexen-1-one, 3-methyl, methylcyclohexenone, caswell no. 561ab, fema no. 3360, 3-methyl-2-cyclohexene-1-one, epa pesticide chemical code 219700, ghl.pd_mitscher_leg0.699
3-methylcyclohex-2-en-1-one
Safety and Handling

Safety and Handling

RTECSNumber : GW7340000

TSCA : Yes

Documents