missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-2-butenal 97.0+%, TCI America™

Manufacturer:  TCI America M07705ML

 View more versions of this product

Catalog No. M07705ML



Specifications

Specifications

3-Methyl-2-butenal
Yellow
5mL
C5H8O
1989
SEPQTYODOKLVSB-UHFFFAOYSA-N
3-methylbut-2-enal
61020
84.12
≥97.0% (GC)
135°C
-20°C
107-86-8
MFCD00010291
3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
CC(=CC=O)C
84.118
CHEBI:15825
Clear Liquid at 20°C

Chemical Identifiers

107-86-8
84.118
SEPQTYODOKLVSB-UHFFFAOYSA-N
61020
3-methylbut-2-enal
C5H8O
MFCD00010291
3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
CHEBI:15825
CC(=CC=O)C
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2461

TSCA : Yes

Recommended Storage : Refrigerator

Documents