missing translation for 'onlineSavingsMsg'
Learn More

3-Methyl-2-butenal 97.0+%, TCI America™

Supplier:  TCI America M077025ML

 View more versions of this product

Catalog No. M077025ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

3-Methyl-2-butenal
-20°C
135°C
MFCD00010291
1989
SEPQTYODOKLVSB-UHFFFAOYSA-N
3-methylbut-2-enal
61020
84.12
Liquid
107-86-8
Yellow
C5H8O
25 mL
3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
CC(=CC=O)C
84.118
CHEBI:15825
≥97.0% (GC)

Chemical Identifiers

107-86-8
84.118
SEPQTYODOKLVSB-UHFFFAOYSA-N
61020
3-methylbut-2-enal
C5H8O
MFCD00010291
3-methyl-2-butenal, prenal, 3-methylcrotonaldehyde, senecialdehyde, 3,3-dimethylacrolein, senecioaldehyde, 2-butenal, 3-methyl, beta,beta-dimethylacrolein, crotonaldehyde, 3-methyl, beta-methylcrotonaldehyde
CHEBI:15825
CC(=CC=O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2461

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions