missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-2-buten-1-ol 98.0+%, TCI America™

Supplier:  TCI America M0714100ML

 View more versions of this product

Catalog No. M0714100ML



Description

Description

Specifications

Specifications

3-Methyl-2-buten-1-ol
Colorless
C5H10O
100 mL
3-methyl-2-buten-1-ol, prenol, 3,3-dimethylallyl alcohol, 2-buten-1-ol, 3-methyl, prenyl alcohol, 3-methyl-2-butenol, dimethylallyl alcohol, 3-methyl-2-butenyl alcohol, 3-methylcrotyl alcohol, 3-methyl-2-butene-1-ol
CC(C)=CCO
86.13
CHEBI:16019
≥98.0% (GC)
556-82-1
140°C
MFCD00002916
1987
ASUAYTHWZCLXAN-UHFFFAOYSA-N
3-methylbut-2-en-1-ol
11173
86.13
Liquid

Chemical Identifiers

556-82-1
86.13
ASUAYTHWZCLXAN-UHFFFAOYSA-N
11173
3-methylbut-2-en-1-ol
C5H10O
MFCD00002916
3-methyl-2-buten-1-ol, prenol, 3,3-dimethylallyl alcohol, 2-buten-1-ol, 3-methyl, prenyl alcohol, 3-methyl-2-butenol, dimethylallyl alcohol, 3-methyl-2-butenyl alcohol, 3-methylcrotyl alcohol, 3-methyl-2-butene-1-ol
CHEBI:16019
CC(C)=CCO
Safety and Handling

Safety and Handling

EINECSNumber : (2)-2393

RTECSNumber : EM9472500

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions