missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-2-butanol 98.0+%, TCI America™

Manufacturer:  TCI America M017125ML

 View more versions of this product

Catalog No. M017125ML



Specifications

Specifications

3-Methyl-2-butanol
Colorless
598-75-4
MFCD00004527
3-methyl-2-butanol, sec-isoamyl alcohol, 2-butanol, 3-methyl, 2-methyl-3-butanol, isopropylmethylcarbinol, methylisopropylcarbinol, 1,2-dimethylpropanol, 1,2-dimethyl-1-propanol, dl-3-methyl-2-butanol, 3-methyl-butan-2-ol
CC(C)C(C)O
88.15
CHEBI:77517
Clear Liquid at 20°C
112°C
25mL
C5H12O
1105
MXLMTQWGSQIYOW-UHFFFAOYSA-N
3-methylbutan-2-ol
11732
88.15
≥98.0% (GC)

Chemical Identifiers

598-75-4
88.15
MXLMTQWGSQIYOW-UHFFFAOYSA-N
11732
3-methylbutan-2-ol
C5H12O
MFCD00004527
3-methyl-2-butanol, sec-isoamyl alcohol, 2-butanol, 3-methyl, 2-methyl-3-butanol, isopropylmethylcarbinol, methylisopropylcarbinol, 1,2-dimethylpropanol, 1,2-dimethyl-1-propanol, dl-3-methyl-2-butanol, 3-methyl-butan-2-ol
CHEBI:77517
CC(C)C(C)O
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0217

TSCA : Yes

Documents