Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-2-butanethiol, 95%, remainder mainly 2-methyl-1-butanethiol

Chemical Identifiers

Chemical Name or Material 3-Methyl-2-butanethiol
CAS 2084-18-6
Molecular Formula C5H12S
Formula Weight 104.22
MDL Number MFCD00039648
UN Number UN3336
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2233006

  SDS
alfa aesar™
B2233006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2233014

  SDS
alfa aesar™
B2233014
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 104.22
Assay Percent Notes remainder mainly 2-methyl-1-butanethiol
Boiling Point 100°C
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P210-P261-P280g-P305+P351+P338-P403+P233
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place.
Keep container tightly closed.
TSCA TSCA
Assay 95%
Health Hazard 2 GHS H Statement
H225-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Flash Point 3°C (37°F)
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: MERCAPTANS, LIQUID, FLAMMABLE, N.O.S.
CAS 2084-18-6
Quantity 5g
Odor Stench
Chemical Name or Material 3-Methyl-2-butanethiol
Molecular Formula C5H12S
MDL Number MFCD00039648
UN Number UN3336
Safety and Handling

Danger

GHS P Statement
P210-P261-P280g-P305+P351+P338-P403+P233
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store in a well-ventilated place.
Keep container tightly closed.

GHS H Statement
H225-H302-H332-H315-H319-H335
Highly flammable liquid and vapour.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.