missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-2-benzothiazolinonehydrazone Hydrochloride, TCI America™

Manufacturer:  TCI America M016025G

 View more versions of this product

Catalog No. M016025G



Specifications

Specifications

3-Methyl-2-benzothiazolinonehydrazone Hydrochloride
25g
C8H10ClN3S
2811
OEZPVSPULCMUQB-DQMXGCRQSA-N
(Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride
9575839
Crystal-Powder at 20°C
Undesignated
4338-98-1
MFCD00005816
MBTH Hydrochloride, Sawicki′s Reagent
CN1C2=CC=CC=C2SC1=NN.Cl
215.699
215.70
≥98.0% (HPLC,T)

Chemical Identifiers

4338-98-1
215.699
OEZPVSPULCMUQB-DQMXGCRQSA-N
9575839
CN1C2=CC=CC=C2SC1=NN.Cl
C8H10ClN3S
MFCD00005816
MBTH Hydrochloride, Sawicki′s Reagent
(Z)-(3-methyl-1,3-benzothiazol-2-ylidene)hydrazine;hydrochloride
Safety and Handling

Safety and Handling

RTECSNumber : DL7160200

TSCA : Yes

Documents