missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-2(5H)-furanone, TCI America™

$123.75 - $1,177.94

Chemical Identifiers

CAS 22122-36-7
Molecular Formula C5H6O2
Molecular Weight (g/mol) 98.101
MDL Number MFCD00191545
InChI Key VGHBEMPMIVEGJP-UHFFFAOYSA-N
Synonym 4-Hydroxy-2-methyl-2-butenoic Acid gamma-Lactone
PubChem CID 30945
IUPAC Name 4-methyl-2H-furan-5-one
SMILES CC1=CCOC1=O
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M10785ML View Documents TCI America
M10785ML
5mL Each for $346.81
 
M107825ML View Documents TCI America
M107825ML
25mL Each for $1,177.94
 
M10781ML View Documents TCI America
M10781ML
1mL Each for $123.75
 
Specifications

Chemical Identifiers

22122-36-7
98.101
VGHBEMPMIVEGJP-UHFFFAOYSA-N
30945
CC1=CCOC1=O
C5H6O2
MFCD00191545
4-Hydroxy-2-methyl-2-butenoic Acid gamma-Lactone
4-methyl-2H-furan-5-one

Specifications

98°C
5mL
C5H6O2
4-Hydroxy-2-methyl-2-butenoic Acid gamma-Lactone
CC1=CCOC1=O
98.101
98.10
≥97.0% (GC)
Yellow
22122-36-7
MFCD00191545
VGHBEMPMIVEGJP-UHFFFAOYSA-N
4-methyl-2H-furan-5-one
30945
Clear Liquid at 20°C
3-Methyl-2(5H)-furanone
Safety and Handling

Safety and Handling

TSCA : No