Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-2(1H)-quinoxalinone, 97%

$60.88 - $219.99

Chemical Identifiers

Chemical Name or Material 3-Methyl-2(1H)-quinoxalinone
CAS 14003-34-0
Molecular Formula C9H8N2O
Formula Weight 160.18
MDL Number MFCD00047567
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5173303

  SDS
alfa aesar™
H5173303
1g Each for $60.88

AAH5173306

  SDS
alfa aesar™
H5173306
5g Each for $219.99
Description & Specifications

Specifications

Formula Weight 160.18
MDL Number MFCD00047567
Melting Point 268° to 273°C
Quantity 1g
Chemical Name or Material 3-Methyl-2(1H)-quinoxalinone
Molecular Formula C9H8N2O
RTECS Number VD3634500
Assay 97%
CAS 14003-34-0