Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1H-pyrazole-1-carboxamide, 98+%

$41.58 - $379.70

Chemical Identifiers

Chemical Name or Material 3-Methyl-1H-pyrazole-1-carboxamide
CAS 873-50-7
Molecular Formula C5H7N3O
Formula Weight 125.13
MDL Number MFCD00020735
Beilstein 115536
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1458406

  SDS
alfa aesar™
A1458406
5g Each for $41.58

AAA1458414

  SDS
alfa aesar™
A1458414
25g Each for $116.16

AAA1458422

  SDS
alfa aesar™
A1458422
100g Each for $379.70
Description & Specifications

Specifications

Formula Weight 125.13
Molecular Formula C5H7N3O
MDL Number MFCD00020735
Quantity 5g
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Assay ≥98%
Melting Point 123° to 127°C
Beilstein 115536
RTECS Number LQ3067000
Health Hazard 1 Warning
Chemical Name or Material 3-Methyl-1H-pyrazole-1-carboxamide
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 873-50-7
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.