missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1H-pyrazole-1-carboxamide, 98+%

Manufacturer:  Alfa Aesar™ A1458406

 View more versions of this product

Catalog No. AAA1458406



Specifications

Specifications

3-Methyl-1H-pyrazole-1-carboxamide
5g
C5H7N3O
115536
PNZZQMXKKNDHNS-UHFFFAOYSA-N
3-methylpyrazole-1-carboxamide
242452
≥98%
123°C to 127°C
873-50-7
MFCD00020735
3-methyl-1h-pyrazole-1-carboxamide, 3-methyl-1-pyrazolecarboxamide, 1h-pyrazole-1-carboxamide, 3 or 5-methyl, 3-methylpyrazolecarboxamide, 1h-pyrazole-1-carboxamide,3-methyl, 3-methyl-1-pyrazolecarboxamide
CC1=NN(C=C1)C(=O)N
125.131
125.13

Chemical Identifiers

873-50-7
125.131
PNZZQMXKKNDHNS-UHFFFAOYSA-N
242452
CC1=NN(C=C1)C(=O)N
C5H7N3O
MFCD00020735
3-methyl-1h-pyrazole-1-carboxamide, 3-methyl-1-pyrazolecarboxamide, 1h-pyrazole-1-carboxamide, 3 or 5-methyl, 3-methylpyrazolecarboxamide, 1h-pyrazole-1-carboxamide,3-methyl, 3-methyl-1-pyrazolecarboxamide
3-methylpyrazole-1-carboxamide
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : LQ3067000

Documents