missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

3-Methyl-1-phenylpyrazole, TCI America™

$67.22 - $226.11

Chemical Identifiers

CAS 1128-54-7
Molecular Formula C10H10N2
Molecular Weight (g/mol) 158.204
MDL Number MFCD00003123
InChI Key RJXLUGSJEMSDPK-UHFFFAOYSA-N
Synonym 3-methyl-1-phenyl-1h-pyrazole, 1-phenyl-3-methylpyrazole, pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, methylphenyl, 1h-pyrazole,3-methyl-1-phenyl, acmc-20an11, ksc494g4n, rjxlugsjemsdpk-uhfffaoysa, 3-methyl-1-phenyl-1h-pyrazole #
PubChem CID 70783
IUPAC Name 3-methyl-1-phenylpyrazole
SMILES CC1=NN(C=C1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M25321G View Documents TCI America
M25321G
1g Each for $67.22
 
M25325G View Documents TCI America
M25325G
5g Each for $226.11
 
Specifications

Chemical Identifiers

1128-54-7
158.204
RJXLUGSJEMSDPK-UHFFFAOYSA-N
70783
CC1=NN(C=C1)C2=CC=CC=C2
C10H10N2
MFCD00003123
3-methyl-1-phenyl-1h-pyrazole, 1-phenyl-3-methylpyrazole, pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, methylphenyl, 1h-pyrazole,3-methyl-1-phenyl, acmc-20an11, ksc494g4n, rjxlugsjemsdpk-uhfffaoysa, 3-methyl-1-phenyl-1h-pyrazole #
3-methyl-1-phenylpyrazole

Specifications

103°C
1g
C10H10N2
3-methyl-1-phenyl-1h-pyrazole, 1-phenyl-3-methylpyrazole, pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, methylphenyl, 1h-pyrazole,3-methyl-1-phenyl, acmc-20an11, ksc494g4n, rjxlugsjemsdpk-uhfffaoysa, 3-methyl-1-phenyl-1h-pyrazole #
CC1=NN(C=C1)C2=CC=CC=C2
158.204
158.20
≥98.0% (GC)
White-Yellow
1128-54-7
MFCD00003123
RJXLUGSJEMSDPK-UHFFFAOYSA-N
3-methyl-1-phenylpyrazole
70783
Crystal-Powder at 20°C
3-Methyl-1-phenylpyrazole
Safety and Handling

Safety and Handling

TSCA : No