missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1-phenyl-2-pyrazolin-5-one, 98+%

Manufacturer:  Alfa Aesar™ A1116136

 View more versions of this product

Catalog No. AAA1116136


 

Description

Description

Applications
3-Methyl-1-phenyl-2-pyrazolin-5-one used as reagent for detection of reducing carbohydrates by ESI/MALDI -MS.

Solubility
Soluble in water(3.3g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Specifications

3-Methyl-1-phenyl-2-pyrazolin-5-one
≥98%
MFCD00003138
14,6713
QELUYTUMUWHWMC-UHFFFAOYSA-N
5-methyl-2-phenyl-4H-pyrazol-3-one
191°C (17mmHg)
4021
500g
Odorless
89-25-8
C10H10N2O
609575
2,4-Dihydro-5-methyl-2-phenyl-3H-pyrazol-3-one; Norphenazone
CC1=NN(C(=O)C1)C2=CC=CC=C2
127°C to 131°C
174.203
CHEBI:31530
174.2

Chemical Identifiers

89-25-8
174.203
QELUYTUMUWHWMC-UHFFFAOYSA-N
4021
C10H10N2O
MFCD00003138
2,4-Dihydro-5-methyl-2-phenyl-3H-pyrazol-3-one; Norphenazone
CHEBI:31530
Safety and Handling

Safety and Handling

RTECSNumber : UQ9625000

TSCA : TSCA

SDS Documents