missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Methyl-1-phenyl-2-phospholene 1-Oxide 95.0+%, TCI America™
$123.03 - $436.19
Chemical Identifiers
| CAS | 707-61-9 |
|---|---|
| Molecular Formula | C11H13OP |
| Molecular Weight (g/mol) | 192.198 |
| MDL Number | MFCD00014518 |
| InChI Key | YMKWWHFRGALXLE-UHFFFAOYSA-N |
| Synonym | 3-methyl-1-phenyl-2-phospholene 1-oxide, 3-methyl-1-phenyl-2-phospholene-1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole 1-oxide, 1h-phosphole, 2,3-dihydro-4-methyl-1-phenyl-, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1h-phosphole 1-oxide, 2-phospholene, 3-methyl-1-phenyl-, 1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1$l^ 5-phosphole 1-oxide, 3-methyl-1-phenyl-4,5-dihydro-1??-phosphol-1-one, 2-phospholene, 1-oxide |
| PubChem CID | 69722 |
| IUPAC Name | 4-methyl-1-phenyl-2,3-dihydro-1$l^{5}-phosphole 1-oxide |
| SMILES | CC1=CP(=O)(CC1)C2=CC=CC=C2 |
Chemical Identifiers
| 707-61-9 | |
| 192.198 | |
| YMKWWHFRGALXLE-UHFFFAOYSA-N | |
| 69722 | |
| CC1=CP(=O)(CC1)C2=CC=CC=C2 |
| C11H13OP | |
| MFCD00014518 | |
| 3-methyl-1-phenyl-2-phospholene 1-oxide, 3-methyl-1-phenyl-2-phospholene-1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole 1-oxide, 1h-phosphole, 2,3-dihydro-4-methyl-1-phenyl-, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1h-phosphole 1-oxide, 2-phospholene, 3-methyl-1-phenyl-, 1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1$l^ 5-phosphole 1-oxide, 3-methyl-1-phenyl-4,5-dihydro-1??-phosphol-1-one, 2-phospholene, 1-oxide | |
| 4-methyl-1-phenyl-2,3-dihydro-1$l^{5}-phosphole 1-oxide |
Specifications
| 707-61-9 | |
| White | |
| C11H13OP | |
| 5 g | |
| YMKWWHFRGALXLE-UHFFFAOYSA-N | |
| 4-methyl-1-phenyl-2,3-dihydro-1$l^{5}-phosphole 1-oxide | |
| 69722 | |
| ≥95.0% (HPLC) | |
| 3-Methyl-1-phenyl-2-phospholene 1-Oxide |
| 64°C | |
| 155°C | |
| MFCD00014518 | |
| 3-methyl-1-phenyl-2-phospholene 1-oxide, 3-methyl-1-phenyl-2-phospholene-1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole 1-oxide, 1h-phosphole, 2,3-dihydro-4-methyl-1-phenyl-, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1h-phosphole 1-oxide, 2-phospholene, 3-methyl-1-phenyl-, 1-oxide, 2,3-dihydro-4-methyl-1-phenyl-1h-phosphole, 1-oxide, 4-methyl-1-phenyl-2,3-dihydro-1$l^ 5-phosphole 1-oxide, 3-methyl-1-phenyl-4,5-dihydro-1??-phosphol-1-one, 2-phospholene, 1-oxide | |
| CC1=CP(=O)(CC1)C2=CC=CC=C2 | |
| 192.198 | |
| 192.20 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-6406
RTECSNumber : SZ6105100
TSCA : Yes