Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1-phenyl-2-phospholene 1-oxide, 97%

$94.04 - $1,219.11

Chemical Identifiers

Chemical Name or Material 3-Methyl-1-phenyl-2-phospholene 1-oxide
CAS 707-61-9
Molecular Formula C11H13OP
Formula Weight 192.19
MDL Number MFCD00014518
Beilstein 609223
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4472606

 
alfa aesar™
4472606
5g Each for $94.04

AA4472614

 
alfa aesar™
4472614
25g Each for $358.65

AA4472622

 
alfa aesar™
4472622
100g Each for $1,219.11
Description & Specifications

Specifications

Physical Form Lumps
Formula Weight 192.19
Chemical Name or Material 3-Methyl-1-phenyl-2-phospholene 1-oxide
TSCA TSCA
Assay 97%
Molecular Formula C11H13OP
MDL Number MFCD00014518
CAS 707-61-9
Beilstein 609223
Color White to cream
Health Hazard 1 Warning
Melting Point 60° to 65°C
Boiling Point 173° to 174°C (0.7mmHg)
Quantity 5g
Flash Point 109°C (228°F)
Odor Unpleasant
Sensitivity Hygroscopic
Health Hazard 2 GHS H Statement
H302
Harmful if swallowed.
Health Hazard 3 GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SZ6105100
Safety and Handling

Warning

GHS P Statement
P264-P270-P301+P312-P330-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Rinse mouth.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302
Harmful if swallowed.