Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1-phenyl-2-butanone, 96%

$114.66 - $114.66

Chemical Identifiers

Chemical Name or Material 3-Methyl-1-phenyl-2-butanone
CAS 2893-05-2
Molecular Formula C11H14O
Formula Weight 162.23
MDL Number MFCD00040487
Beilstein 2042880
Synonym Benzyl isopropyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0717606

  SDS
alfa aesar™
L0717606
5g Each for $114.66
Description & Specifications

Specifications

Formula Weight 162.23
Chemical Name or Material 3-Methyl-1-phenyl-2-butanone
CAS 2893-05-2
Assay 96%
Molecular Formula C11H14O
MDL Number MFCD00040487
Beilstein 2042880
Synonym Benzyl isopropyl ketone
Boiling Point 237° to 240°C
Quantity 5g

Applications
3-Methyl-1-phenyl-2-butanone is used as pharmaceutical intermediate.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.