missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 3-Methyl-1-phenyl-1H-pyrazole, 98%

Manufacturer:  Alfa Aesar™ A1484109

 View more versions of this product

Catalog No. AAA1484109



Description

Description

Applications
3-Methyl-1-phenyl-1H-pyrazole is used to produce 3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde at the temperature of 90-100°C with reagent phosphorus oxychloride.

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

3-Methyl-1-phenyl-1H-pyrazole
Odorless
10g
C10H10N2
117870
RJXLUGSJEMSDPK-UHFFFAOYSA-N
3-methyl-1-phenylpyrazole
70783
98%
>110°C (230°F)
35°C to 38°C
1128-54-7
MFCD00003123
3-methyl-1-phenyl-1h-pyrazole, 1-phenyl-3-methylpyrazole, pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, methylphenyl, 1h-pyrazole,3-methyl-1-phenyl, acmc-20an11, ksc494g4n, rjxlugsjemsdpk-uhfffaoysa, 3-methyl-1-phenyl-1h-pyrazole #
CC1=NN(C=C1)C2=CC=CC=C2
158.204
158.2

Chemical Identifiers

1128-54-7
158.204
RJXLUGSJEMSDPK-UHFFFAOYSA-N
70783
CC1=NN(C=C1)C2=CC=CC=C2
C10H10N2
MFCD00003123
3-methyl-1-phenyl-1h-pyrazole, 1-phenyl-3-methylpyrazole, pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, 3-methyl-1-phenyl, 1h-pyrazole, methylphenyl, 1h-pyrazole,3-methyl-1-phenyl, acmc-20an11, ksc494g4n, rjxlugsjemsdpk-uhfffaoysa, 3-methyl-1-phenyl-1h-pyrazole #
3-methyl-1-phenylpyrazole
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents