Please login to your online account to display your discounted pricing

3-Methyl-1-pentyn-3-ol 99+%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 3-Methyl-1-pentyn-3-ol
Name Note 99+%
CAS 77-75-8
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C6H10O
Linear Formula C2H5C(CH3)(OH)Cidentical toCH
Formula Weight 98.14
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC160065000

  SDS
thermo fisher scientific
160065000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Liquid or Low Melting Solid
Quantity 500g
Color Clear slightly yellow
Merck Index 15, 5913
RTECS Number SC4900000
Chemical Name or Material 3-Methyl-1-pentyn-3-ol
MDL Number MFCD00004479
Health Hazard 1 GHS Signal Word: Warning
Molecular Formula C6H10O
TSCA TSCA
Synonym Meparfynol
EINECS Number 201-055-5
CAS Min % 98.0
Linear Formula C2H5C(CH3)(OH)Cidentical toCH
Packaging Glass bottle
Infrared Spectrum Authentic
Formula Weight 98.14
CAS 77-75-8
Assay Percent Range 98+%
Density 0.8600g/mL
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Refractive Index 1.4300 to 1.4320 (20°C, 589nm)
Name Note 99+%
CAS Max % 100.0
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
Flammable liquid and vapor.